missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Mycoalert™ Plus Mycoplasma Detection Kit

Product Code. 13459219
Change view
Click to view available options
Quantity:
30 Tests
Unit Size:
30 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13459219 30 Tests 30 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13459219 Supplier Lonza™ Supplier No. LT07703
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Content And Storage Mycoalert plus reagent: 3 x 1.2mL, Mycoalert plus assay buffer: 3 x 3mL, Mycoalert substrate: 3 x 1.2mL
2 to 8°C
Cell Type Mammalian Cells
Product Type Mycoplasma Detection
Quantity 30 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.