missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYCNOS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103497
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MYCNOS.
This gene is transcribed in antisense to the v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog gene (MYCN). It is thought to encode a small, novel protein that stabilizes MYCN, prevents apoptosis, and promotes cell proliferation. Transcripts at this locus may also act directly as functional RNAs to recruit transcriptional regulators to the promoter of MYCN and stimulate transcription of this oncogene. The MYCNOS gene therefore functions through both RNA and protein products.
Specifications
| MYCNOS | |
| Polyclonal | |
| Unconjugated | |
| MYCNOS | |
| MYCN opposite strand; MYCN-AS1; MYCNOS; NCYM; N-CYM; NYCM | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10408, 110714 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P40205 | |
| MYCNOS | |
| A synthesized peptide derived from human MYCNOS(Accession P40205), corresponding to amino acid residues R73-K109. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction