missing translation for 'onlineSavingsMsg'
Learn More

MYCL1/L-Myc Antibody (PCRP-MYCL-2D5), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18341865 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341865 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341865 Supplier Novus Biologicals Supplier No. NBP314011JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MYCL1/L-Myc Monoclonal antibody specifically detects MYCL1/L-Myc in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYCL1/L-Myc
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-MYCL-2D5
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias BHLHE38, bHLHe38myc-related gene from lung cancer, Class E basic helix-loop-helix protein 38, LMYCl-myc-1 proto-oncogene, MYCL, protein L-Myc-1, v-myc avian myelocytomatosis viral oncogene homolog 1, lung carcinoma derived, v-myc myelocytomatosis viral oncogene homolog 1, lung carcinoma derived (avian)
Host Species Mouse
Immunogen Recombinant full-length human MYCL1/L-Myc protein (Uniprot: P12524)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4610
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.