missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYCL1/L-Myc Antibody (PCRP-MYCL-2D5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14011CL1
This item is not returnable.
View return policy
Description
MYCL1/L-Myc Monoclonal antibody specifically detects MYCL1/L-Myc in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Western Blot,Protein Array,Immunocytochemistry/Immunofluorescence
Specifications
| MYCL1/L-Myc | |
| Monoclonal | |
| CoraFluor 1 | |
| BHLHE38, bHLHe38myc-related gene from lung cancer, Class E basic helix-loop-helix protein 38, LMYCl-myc-1 proto-oncogene, MYCL, protein L-Myc-1, v-myc avian myelocytomatosis viral oncogene homolog 1, lung carcinoma derived, v-myc myelocytomatosis viral oncogene homolog 1, lung carcinoma derived (avian) | |
| Recombinant full-length human MYCL1/L-Myc protein (Uniprot: P12524) | |
| 0.1 mL | |
| Breast Cancer | |
| 4610 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry, ELISA, Immunoprecipitation, Western Blot, Protein Array | |
| PCRP-MYCL-2D5 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction