missing translation for 'onlineSavingsMsg'
Learn More

MYCL1/L-Myc Antibody (PCRP-MYCL-2D5) - Azide and BSA Free, Novus Biologicals™

Product Code. 18396502
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396502 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396502 Supplier Bio-Techne Supplier No. NBP314011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MYCL1/L-Myc Monoclonal antibody specifically detects MYCL1/L-Myc in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Protein Array
TRUSTED_SUSTAINABILITY

Spécification

Antigen MYCL1/L-Myc
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array
Classification Monoclonal
Clone PCRP-MYCL-2D5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg /million cells, ELISA, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunoprecipitation 1 to 2 μg per 100-500 μg protein, Protein Array
Formulation 10mM PBS without Preservative
Gene Alias BHLHE38, bHLHe38myc-related gene from lung cancer, Class E basic helix-loop-helix protein 38, LMYCl-myc-1 proto-oncogene, MYCL, protein L-Myc-1, v-myc avian myelocytomatosis viral oncogene homolog 1, lung carcinoma derived, v-myc myelocytomatosis viral oncogene homolog 1, lung carcinoma derived (avian)
Host Species Mouse
Immunogen Recombinant full-length human MYCL1/L-Myc protein (Uniprot: P12524)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4610
Target Species Human
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.