missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYCL1/L-Myc Antibody (PCRP-MYCL-2D5), Allophycocyanin, Novus Biologicals™
Shop All Bio Techne ProductsDescription
MYCL1/L-Myc Monoclonal antibody specifically detects MYCL1/L-Myc in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | MYCL1/L-Myc |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | PCRP-MYCL-2D5 |
| Conjugate | APC |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | BHLHE38, bHLHe38myc-related gene from lung cancer, Class E basic helix-loop-helix protein 38, LMYCl-myc-1 proto-oncogene, MYCL, protein L-Myc-1, v-myc avian myelocytomatosis viral oncogene homolog 1, lung carcinoma derived, v-myc myelocytomatosis viral oncogene homolog 1, lung carcinoma derived (avian) |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human MYCL1/L-Myc protein (Uniprot: P12524) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?