missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYCBPAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570407
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 92%; Dog: 92%; Guinea Pig: 85%; Horse: 77%; Human: 100%; Mouse: 92%; Pig: 92%; Rabbit: 85%; Rat: 92%.
MYCBPAP is a protein coding gene.
Specifications
| MYCBPAP | |
| Polyclonal | |
| Unconjugated | |
| MYCBPAP | |
| 4932408B01Rik; AMAM-1; AMAP1; AMAP-1; AMY1-associating protein 1; AMY-1-binding protein 1; AW125474; cochlea derived protein of 106 kDa; COD106; LOW QUALITY PROTEIN: MYCBP-associated protein; MYCBP associated protein; MYCBPAP; mycbpap {ECO:0000312; MYCBP-associated protein; MYCBP-associated protein; LOW QUALITY PROTEIN: MYCBP-associated protein; Organ of Corti membrane-associated protein of 106 kDa; RGD:1359123}; testis secretory sperm-binding protein Li 214e | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84073 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TBZ2 | |
| MYCBPAP | |
| synthetic peptide directed towards the N-terminal region of human MYCBPAP. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction