Learn More
Myc Tag Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1981
276.80 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-981 detects c-Myc tagged proteins. PA1-981 has been successfully used in WB and immunofluorescence procedures to detect the presence of expressed c-Myc tagged proteins in transfected mammalian cells. PA1-981 immunogen is a synthetic peptide corresponding to residues C E(410) Q K L I S E E D L(419) of human c-Myc. PA1-981 immunizing peptide (PEP-035) is available for use in neutralization and control experiments.
Myc is a proto-oncogene and encodes a nuclear phosphoprotein that plays a role in cell cycle progression, apoptosis and cellular transformation. The encoded protein forms a heterodimer with the related transcription factor MAX. This complex binds to the E box DNA consensus sequence and regulates the transcription of specific target genes. Amplification of this gene is frequently observed in numerous human cancers. Translocations involving this gene are associated with Burkitt lymphoma and multiple myeloma in human patients. There is evidence to show that translation initiates both from an upstream, in-frame non-AUG (CUG) and a downstream AUG start site, resulting in the production of two isoforms with distinct N-termini.Specifications
Myc Tag | |
Polyclonal | |
Unconjugated | |
MYC | |
46-0603; AU016757; avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; class E basic helix-loop-helix protein 39; cmyc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; MRTL; MYC; Myc protein; Myc proto-oncogene protein; Myc Tag; Myc2; MYCC; myc-related translation/localization regulatory factor; myelocytomatosis oncogene; Niard; Nird; Proto-oncogene c-Myc; R95025; R950-25; transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4609 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
ChIP Assay, Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P01106 | |
MYC | |
Synthetic peptide corresponding to residues C E(410) Q K L I S E E D L(419) of human c-Myc. | |
100 μL | |
Primary | |
Tag | |
Antibody | |
IgG |