Learn More
Myc Tag Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Brand: Invitrogen A21281
417.60 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Description
Myc is a proto-oncogene and encodes a nuclear phosphoprotein that plays a role in cell cycle progression, apoptosis and cellular transformation. The encoded protein forms a heterodimer with the related transcription factor MAX. This complex binds to the E box DNA consensus sequence and regulates the transcription of specific target genes. Amplification of this gene is frequently observed in numerous human cancers. Translocations involving this gene are associated with Burkitt lymphoma and multiple myeloma in human patients. There is evidence to show that translation initiates both from an upstream, in-frame non-AUG (CUG) and a downstream AUG start site, resulting in the production of two isoforms with distinct N-termini.Order Info
Shipping Condition: Wet Ice
Specifications
Myc Tag | |
Polyclonal | |
Unconjugated | |
MYC | |
46-0603; AU016757; avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; class E basic helix-loop-helix protein 39; cmyc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; MRTL; MYC; Myc protein; Myc proto-oncogene protein; Myc Tag; Myc2; MYCC; myc-related translation/localization regulatory factor; myelocytomatosis oncogene; Niard; Nird; Proto-oncogene c-Myc; R95025; R950-25; transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog | |
Chicken | |
purified | |
RUO | |
4609 | |
4° C | |
Liquid | |
Tag |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
P01106 | |
MYC | |
Was raised against the peptide EQKLISEEDL. | |
100 μL | |
Primary | |
Tag | |
Antibody | |
IgY |
For Research Use Only. Not for use in diagnostic procedures.