missing translation for 'onlineSavingsMsg'
Learn More

Myc Tag Polyclonal Antibody, HRP, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA5141013

Additional Details : Weight : 0.01000kg

Product Code. 17896572

  • £383.00 / 100µL
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Myc is a proto-oncogene and encodes a nuclear phosphoprotein that plays a role in cell cycle progression, apoptosis and cellular transformation. The encoded protein forms a heterodimer with the related transcription factor MAX. This complex binds to the E box DNA consensus sequence and regulates the transcription of specific target genes. Amplification of this gene is frequently observed in numerous human cancers. Translocations involving this gene are associated with Burkitt lymphoma and multiple myeloma in human patients. There is evidence to show that translation initiates both from an upstream, in-frame non-AUG (CUG) and a downstream AUG start site, resulting in the production of two isoforms with distinct N-termini.
Specifications

Specifications

Myc Tag
Polyclonal
HRP
MYC
46-0603; AU016757; avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; class E basic helix-loop-helix protein 39; cmyc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; MRTL; MYC; Myc protein; Myc proto-oncogene protein; Myc Tag; Myc2; MYCC; myc-related translation/localization regulatory factor; myelocytomatosis oncogene; Niard; Nird; Proto-oncogene c-Myc; R95025; R950-25; transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog
Rabbit
Antigen affinity chromatography
RUO
4609
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Western Blot
1 mg/mL
PBS with 0.2% BSA, 40% glycerol and no preservative; pH 7.4
P01106
MYC
Synthetic peptide within human Myc aa 410-420.
100 μL
Primary
Human, Tag
Liquid
Tag
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Product Certifications
Offerte speciali

Offerte speciali