missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myc Tag Monoclonal Antibody (Myc.A7), HRP, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA121316HRP
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA1-21316-HRP has been successfully used in Western blotting applications on myc tagged samples.
Myc is a proto-oncogene and encodes a nuclear phosphoprotein that plays a role in cell cycle progression, apoptosis and cellular transformation. The encoded protein forms a heterodimer with the related transcription factor MAX. This complex binds to the E box DNA consensus sequence and regulates the transcription of specific target genes. Amplification of this gene is frequently observed in numerous human cancers. Translocations involving this gene are associated with Burkitt lymphoma and multiple myeloma in human patients. There is evidence to show that translation initiates both from an upstream, in-frame non-AUG (CUG) and a downstream AUG start site, resulting in the production of two isoforms with distinct N-termini.Specifications
Myc Tag | |
Monoclonal | |
1 mg/mL | |
antibody with 0.07% Kathon | |
P01106 | |
MYC | |
Synthetic peptide: EQKLISEEDL (Myc) | |
50 μL | |
Primary | |
Tag | |
Antibody | |
IgG1 |
Western Blot | |
Myc.A7 | |
HRP | |
MYC | |
46-0603; AU016757; avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; class E basic helix-loop-helix protein 39; cmyc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; MRTL; MYC; Myc protein; Myc proto-oncogene protein; Myc Tag; Myc2; MYCC; myc-related translation/localization regulatory factor; myelocytomatosis oncogene; Niard; Nird; Proto-oncogene c-Myc; R95025; R950-25; transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog | |
Mouse | |
Protein A | |
RUO | |
4609 | |
4° C | |
Liquid | |
Tag |