Learn More
Myc Tag Monoclonal Antibody (Myc.A7), DyLight™ 800 4X PEG, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA121316D800
Code : NEW
Additional Details : Weight : 0.00900kg
Description
MA1-21316-D800 detects Myc tag from various samples. MA1-21316-D800 has been successfully used for fluorescent detection of Myc tagged proteins in Western Blotting. DyLight 800 4X PEG has an excitation/emission of 783/797 nm.
Myc Tag Monoclonal antibody specifically detects Myc Tag in Tag samples. It is validated for Western Blot.Specifications
Myc Tag | |
Monoclonal | |
1 mg/mL | |
MES with proprietary stabilizer and no preservative | |
P01106 | |
MYC | |
EQKLISEEDL synthetic peptide conjugated to KLH | |
50 μL | |
Primary | |
4° C, do not freeze | |
Liquid | |
Tag |
Western Blot | |
Myc.A7 | |
DyLight 800 4X PEG | |
MYC | |
46-0603; AU016757; avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; class E basic helix-loop-helix protein 39; cmyc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; MRTL; MYC; Myc protein; Myc proto-oncogene protein; Myc Tag; Myc2; MYCC; myc-related translation/localization regulatory factor; myelocytomatosis oncogene; Niard; Nird; Proto-oncogene c-Myc; R95025; R950-25; transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog | |
Mouse | |
Protein A | |
RUO | |
4609 | |
Antibody | |
IgG1 |