missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYBPC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571701
This item is not returnable.
View return policy
Description
MYBPC3 Polyclonal Antibody for IHC (P), Western Blot
Myosin binding protein C, cardiac-type (MYBPC3) is a thick filament-associated protein located in the crossbridge region of vertebrate striated muscle a bands. In vitro it binds MHC, F-actin and native thin filaments. It modifies the activity of actin-activated myosin ATPase. It may modulate muscle contraction or may play a more structural role. Mutations affecting the gene can result in cardiomyopathy - familial hypertrophic 4, cardiomyopathy - dilated 1MM, or left ventricular non-compaction 10.
Specifications
| MYBPC3 | |
| Polyclonal | |
| Unconjugated | |
| MYBPC3 | |
| cardiac C-protein; cardiac MyBP-C; cardiac myosin binding protein C; CMD1MM; CMH4; C-protein, cardiac muscle isoform; DKFZp779E1762; FHC; LVNC10; MYBP-C; Mybpc3; myosin binding protein C, cardiac; myosin binding protein C3; myosin-binding protein C, cardiac-type; truncated cardiac myosin-binding protein C | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 17868, 295929, 4607 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O70468, P56741, Q14896 | |
| MYBPC3 | |
| KLH conjugated synthetic peptide between 189-218 amino acids from the N-terminal region of human MYBPC3. | |
| 400 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction