missing translation for 'onlineSavingsMsg'
Learn More

MyBPC3 Antibody [DyLight 680], Novus Biologicals™

Product Code. 30735638 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735638 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735638 Supplier Novus Biologicals Supplier No. AF7439Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

MyBPC3 Polyclonal antibody specifically detects MyBPC3 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MyBPC3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias Cardiac MyBP-C, CMH4, C-protein, cardiac muscle isoform, DKFZp779E1762, FHC, MYBP-C, myosin binding protein C, cardiac, myosin-binding protein C, cardiac, myosin-binding protein C, cardiac-type
Host Species Sheep
Immunogen E. coli-derived recombinant human MyBPC3, Arg1001-Lys1103, Accession # Q14896
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4607
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.