Learn More
Invitrogen™ MYBPC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589164
Description
Immunogen sequence: LSVQIENMKD TATIRIRVVE KAGPPINVMV KEVWGTNALV EWQAPKDDGN SEIMGYFVQK ADKKTMEWFN VYERNRHTSC TVSDLIVGNE YYFRVYTENI CGLSDSPGVS KNTARILKTG ITFKPFEYKE HDFRMAPKFL TPLIDRVVVA GYSAALNCAV RGHPKPKVVW MKNKMEIRED PKFLITNYQG VLTLNIRRPS PFDAGTYTCR AVNELGEALA ECKLEVRVPQ; Positive Samples: Mouse skeletal muscle, Rat skeletal muscle.
The gene MYBPC2 belongs to the myosin-binding protein C family, which consists of fast-, slow-, and cardiac-type isoforms. These isoforms are myosin-associated proteins that are located in the C region of A bands in striated muscle. The protein encoded by MYBPC2 is the fast-type isoform. Mutations in genes encoding the slow-type and cardiac-type isoforms have been linked to distal arthrogryposis, type 1, and hypertrophic cardiomyopathy, respectively. MYBPC2-related diseases include Lethal Congenital Contracture Syndrome 4 and Nemaline Myopathy 9.
Specifications
| MYBPC2 | |
| Polyclonal | |
| Unconjugated | |
| MYBPC2 | |
| C-protein, skeletal muscle fast isoform; Fast MyBP-C; fast-type C-protein; fast-type muscle myosin-binding-protein C; MYBPC; Mybpc2; MYBPCF; myosin binding protein C, fast type; myosin binding protein C, fast-type; myosin-binding protein C, fast-type; mypc2; testicular tissue protein Li 126 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 233199, 292879, 4606 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.82 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14324, Q5XKE0 | |
| MYBPC2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 912-1141 of human MYBPC2 (NP_0045243). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.