missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calretinin Antibody (rCALB2/7123) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20777
This item is not returnable.
View return policy
Description
Calretinin Monoclonal antibody specifically detects Calretinin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Calretinin | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| 29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR | |
| Recombinant human Calretinin protein fragment (around aa23-242) (exact sequence is proprietary) | |
| 100 μg | |
| Cellular Markers, Phospho Specific | |
| 794 | |
| Store at -20°C to -70°C. Avoid freeze/thaw cycles. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rCALB2/7123 | |
| Unconjugated | |
| 10mM PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction