missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYBPC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101687
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MYBPC1.
Myosin binding protein C (MYBPC) is a component of the thick filament of striated muscle, with the slow-type isoform designated MYBPC1. Both the fast-type (MYBPC2) and slow-type MYBPC protein contains seven immunoglobulin C2 motifs and three fibronectin type-III repeats. Multiple isoforms of MYBPC1 are known to exist, and are present in varying amounts in different skeletal muscles. It is thought that the MYBPC1 slow subfamily may play important roles in the assembly and stabilization of sarcomeric M- and A-bands and regulate the contractile properties of the actomyosin filaments.
Specifications
| MYBPC1 | |
| Polyclonal | |
| Unconjugated | |
| MYBPC1 | |
| 8030451F13Rik; C-protein, skeletal muscle slow isoform; LCCS4; muscle C-protein; MyBP-C slow; Mybpc1; MYBPCC; MYBPCS; myosin binding protein C, slow type; myosin binding protein C, slow-type; myosin-binding protein C, slow-type; skeletal muscle C-protein; slow MyBP-C; Slow-type C-protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 109272, 362867, 4604 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q00872, Q63518 | |
| MYBPC1 | |
| A synthesized peptide derived from human MYBPC1(Accession Q00872), corresponding to amino acid residues A217-G267. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction