missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYBPC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-86385
This item is not returnable.
View return policy
Description
MYBPC1 Polyclonal specifically detects MYBPC1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MYBPC1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q00872 | |
| MYBPC1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:YYSQPILVKEIIEPPKIRIPRHLKQTYIRRVGEAVNLVIPFQGKPRPELTWKKDGAEIDKNQINIRNSETDTIIFIRKAERSHSGKYDLQVKVDKFVETASIDIQIIDRPGPPQIVKIEDVWGENVALTWTPPKDDGNAAIT | |
| 0.1 mL | |
| Cellular Signaling | |
| 4604 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| C-protein, skeletal muscle slow isoform, MYBPCC, MYBPCS, myosin binding protein C, slow type, myosin-binding protein C, slow-type, skeletal muscle C-protein, Slow MyBP-C, slow-type | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction