missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYBPC1 Antibody (83B24) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50295
This item is not returnable.
View return policy
Description
MYBPC1 Monoclonal antibody specifically detects MYBPC1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MYBPC1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000 | |
| C-protein, skeletal muscle slow isoform, MYBPCC, MYBPCS, myosin binding protein C, slow type, myosin-binding protein C, slow-type, skeletal muscle C-protein, Slow MyBP-C, slow-type | |
| Crude preparation of human skeletal myosin | |
| 0.1 mL | |
| Cellular Signaling | |
| 4604 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| 83B24 | |
| Unconjugated | |
| Ascites | |
| Mouse | |
| Ascites | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction