missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myb Polyclonal Antibody

Myb Polyclonal Antibody
Brand: Invitrogen PA518744
This item is not returnable.
View return policy
Description
Myb Polyclonal Antibody for Western Blot, IHC
Specifications
| Myb | |
| Polyclonal | |
| Unconjugated | |
| MYB | |
| AI550390; Cmyb; c-myb; cmyb protein; c-myb_CDS; c-myb10A_CDS; c-myb13A_CDS; c-myb14A_CDS; c-myb8B_CDS; efg; gag-myb protein; M16449; MYB; myb proto-oncogene protein; MYB proto-oncogene, transcription factor; Mybl2; myb-related protein B; myeloblastosis oncogene; myeloblastosis oncogene-like 2; myeloblastosis proto-oncogene product; oncogene AMV; proto-oncogene c-Myb; RP1-32B1.1; transcription factor cmyb; transcriptional activator Myb; tumor-specific myb protein; v-myb avian myeloblastosis viral oncogene homolog | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 4602 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P10242 | |
| MYB | |
| Synthetic peptide sequence (QRHYNDEDPEKEKR) corresponding to the internal amino acids of MYB (aa 281-295). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction