missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYADM Mouse anti-Human, Clone: MYADM/971, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14420.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against full length recombinant human MYADM.
- High specificity
- High reactivity
Specifications
| MYADM | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Q96S97 | |
| MYADM | |
| Recombinant protein corresponding to full length human MYADM. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| MYADM/971 | |
| Mouse monoclonal antibody raised against full length recombinant human MYADM. | |
| In PBS (0.05% BSA, 0.05% sodium azide). | |
| SB135 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 91663 | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction