Learn More
Restek MXT-1 Cap. Column 60m 0.28mm ID 1.00um Coil - 6" Diameter without cage (6.0"ID/6.25"OD)
Brand:  Restek 70157-276
Encompass_Preferred
MXT-1 Cap. Column 60m 0.28mm ID 1.00um Coil - 6" Diameter without cage (6.0"ID/6.25"OD)
Product Code. 16639206
  • £1575.42 / Each