missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MXI1 Polyclonal Antibody

MXI1 Polyclonal Antibody
Brand: Invitrogen PA593131
This item is not returnable.
View return policy
Description
MXI1 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| MXI1 | |
| Polyclonal | |
| Unconjugated | |
| Mxi1 | |
| bHLHc11; class C basic helix-loop-helix protein 11; ENSMUSG00000067085; Gm10197; Mad2; MAX dimerization protein 2; Max interacting protein 1; MAX interactor 1; MAX interactor 1, dimerization protein; max-interacting protein 1; Max-interacting protein 1-like protein; Max-related transcription factor; MGC43220; MXD2; MXI; MXI1; MXI-WR; RP11-549L6.1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17859, 25701, 4601 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.61 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O09015, P50539, P50540 | |
| Mxi1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 76-295 of human MXI1 (NP_5691572). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction