missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MXI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568756
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 86%; Mouse: 100%; Pig: 100%; Rat: 100%.
It is now well established that Myc regulation of cell proliferation and differentiation involves a family of related transcription factors. One such factor, Max, is an obligate heterodimeric partner for Myc and can also form heterodimers with at least four related proteins designated Mad 1, Mxi1, Mad 3 and Mad 4. Like Mad 1 and Mxi1, association of Mad 3 and Mad 4 with Max results in transcriptional repression. Both Myc and the Mad proteins have short half-lives and their synthesis is tightly regulated, while Max expression is constitutive and relatively stable. Two related mammalian cDNAs have been identified and shown to encode Mad-binding proteins. Both possess sequence homology with the yeast transcription repressor Sin3 including four conserved paired amphipathic helix (PAH) domains. mSin3A and mSin3B specifically interact with the Mad proteins via their second paired amphipathic helix domain (PAH2). It has been suggested that Mad-Max heterodimers repress transcription by tethering mSin3 to DNA as corepressors.
Specifications
| MXI1 | |
| Polyclonal | |
| Unconjugated | |
| Mxi1 | |
| bHLHc11; class C basic helix-loop-helix protein 11; ENSMUSG00000067085; Gm10197; Mad2; MAX dimerization protein 2; Max interacting protein 1; MAX interactor 1; MAX interactor 1, dimerization protein; max-interacting protein 1; Max-interacting protein 1-like protein; Max-related transcription factor; MGC43220; MXD2; MXI; MXI1; MXI-WR; RP11-549L6.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17859, 4601 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P50539, P50540 | |
| Mxi1 | |
| synthetic peptide directed towards the N-terminal region of mouse MXI1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction