missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MXD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19547-1-AP-150UL
This item is not returnable.
View return policy
Description
MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor.Specifications
| MXD1 | |
| Polyclonal | |
| Unconjugated | |
| MXD1 | |
| MAD, MAD1, MAX dimerization protein 1, Max dimerizer 1, MXD1, Protein MAD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17119, 362391, 4084 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P50538, Q05195 | |
| Mxd1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction