missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MX1 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 82677-1-RR-100UL
This item is not returnable.
View return policy
Description
The MX1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human MX1. This antibody recognizes human antigen. The MX1 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| MX1 | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| IFI-78K; IFI78; MX; MxA | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human | |
| 75 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 2L19 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 4599 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur