missing translation for 'onlineSavingsMsg'
Learn More

MW-150, MedChemExpress

Product Code. 16427667
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16427667 5 mg 5mg
16425696 100 mg 100mg
16404918 10 mg 10mg
16401167 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16427667 Supplier Medchem Express Supplier No. HY1201115mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MW150 (MW01-18-150SRM) is a selective, CNS penetrant, and orally active inhibitor of p38α MAPK with a Ki of 101 nM. MW-150 inhibits the ability of the endogenous p38α MAPK to phosphorylate an endogenous substrate MK2 in activated glia.

MW150 (MW01-18-150SRM) is a selective, CNS penetrant, and orally active inhibitor of p38α MAPK with a Ki of 101 nM. MW-150 inhibits the ability of the endogenous p38α MAPK to phosphorylate an endogenous substrate MK2 in activated glia.

Specifications

Chemical Name or Material MW-150
Physical Form Solid
CAS 1628502-91-9
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C24H23N5
For Use With (Application) Neuroscience-Neuromodulation
Synonym MW01-18-150SRM
Solubility Information DMSO : 40 mg/mL (104.86 mM; Need ultrasonic)
SMILES CN1CCN(C2=NN=C(C3=CC=C4C=CC=CC4=C3)C(C5=CC=NC=C5)=C2)CC1
Molecular Weight (g/mol) 381.47
Formula Weight 381.47
Percent Purity 98.01%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.