missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MVP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527212
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa, NIH-3T3. Predicted reactivity: Mouse (91%), Rat (91%), Dog (96%), Cat (95%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the major vault protein which is a lung resistance-related protein. Vaults are multi-subunit structures that may be involved in nucleo-cytoplasmic transport. This protein mediates drug resistance, perhaps via a transport process. It is widely distributed in normal tissues, and overexpressed in multidrug-resistant cancer cells. The protein overexpression is a potentially useful marker of clinical drug resistance. This gene produces two transcripts by using two alternative exon 2 sequences; however, the open reading frames are the same in both transcripts.
Specifications
| MVP | |
| Polyclonal | |
| Unconjugated | |
| MVP | |
| 2310009M24Rik; LRP; Lung resistance-related protein; major vault protein; MVP; MVP1; Testicular Secretory Protein Li 30; VAULT1; vRNP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 78388, 9961 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q14764, Q9EQK5 | |
| MVP | |
| Recombinant protein encompassing a sequence within the C-terminus region of human MVP/LRP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction