missing translation for 'onlineSavingsMsg'
Learn More

MVP Antibody (1014) - Azide and BSA Free, Novus Biologicals™

Product Code. 18342573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342573 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342573 Supplier Novus Biologicals Supplier No. NBP311555

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MVP Monoclonal antibody specifically detects MVP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MVP
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 1014
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunocytochemistry/Immunofluorescence
Formulation 10 mM PBS
Gene Alias LRPVAULT1, Lung resistance-related protein, major vault protein
Host Species Mouse
Immunogen proteins precipitated from human breast cancer MCF-7 cells
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9961
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.