Learn More
Invitrogen™ MVK Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592415
Beschreibung
Immunogen sequence: RLQPHSNGKV DLSLPNIGIK RAWDVARLQS LDTSFLEQGD VTTPTSEQVE KLKEVAGLPD DCAVTERLAV LAFLYLYLSI CRKQRALPSL DIVVWSELPP GAGLGSSAAY SVCLAAALLT VCEEIPNPLK DGDCVNRWTK EDLELINKWA FQGERMIHGN PSGVDNAVST WGGALRYHQG KISSLKRSPA LQILLTNTKV PRNTRALVAG VRNRLLKFPE IVAPLLTSID AISLECERVL GEMGEAPAPE QYLVLEELID M; Positive Samples: Raji, BT-474; Cellular Location: Cytoplasm.
Mevalonate kinase (MVK) is an early enzyme in isoprenoid and sterol synthesis. Mevalonate kinase catalyzes the ATP-dependent phosphorylation of mevalonic acid to form mevalonate 5-phosphate. Deficiency in MVK activity contributes to mevalonic aciduria and hyperimmunoglobulinemia D/periodic fever syndrome (HIDS).
Spezifikation
| MVK | |
| Polyclonal | |
| Unconjugated | |
| Mvk | |
| 2310010A05Rik; AI256848; AI414037; KIME; LH receptor mRNA-binding protein; LHR mRNA binding protein; Lrbp; Mevalonate kinase; mevalonate kinase 1; MK; Mvk; MVLK; POROK3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4598 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.72 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q03426 | |
| Mvk | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-300 of human MVK (NP_0004221). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.