missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MVK Antibody (OTI 1D7), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72837AF594
This item is not returnable.
View return policy
Description
MVK Monoclonal specifically detects MVK in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| MVK | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MVK | |
| Full length recombinant protein of human MVK (NP_000422) produced in HEK293T cells. | |
| metabolism | |
| 4598 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI 1D7 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| EC 2.7.1.36, LH receptor mRNA-binding protein, LRBP, mevalonate kinase, mevalonate kinase (mevalonic aciduria), mevalonate kinase 1, MKFLJ96772, MVLK | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction