missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MVD Recombinant Rabbit Monoclonal Antibody (7G4B9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542627
This item is not returnable.
View return policy
Description
Positive test controls include: HepG2, Rat brain. Immunogen sequence: PSFAQLTMKD SNQFHATCLD TFPPISYLNA ISWRIIHLVH RFNAHHGDTK VAYTFDAGPN AVIFTLDDTV AEFVAAVWHG FPPGSNGDTF LKGLQVRPAP L.
The enzyme mevalonate pyrophosphate decarboxylase catalyzes the conversion of mevalonate pyrophosphate into isopentenyl pyrophosphate in one of the early steps in cholesterol biosynthesis. It decarboxylates and dehydrates its substrate while hydrolyzing ATP.
Specifications
| MVD | |
| Recombinant Monoclonal | |
| 2.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P53602, Q62967 | |
| Mvd | |
| A synthetic peptide corresponding to a sequence within amino acids 250-350 of human MVD (P53602). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 7G4B9 | |
| Unconjugated | |
| Mvd | |
| C78718; diphosphomevalonate decarboxylase; FP17780; MDDase; mevalonate (diphospho) decarboxylase; Mevalonate (diphospho)decarboxylase; mevalonate diphosphate decarboxylase; mevalonate pyrophosphate decarboxylase; Mpd; MVD; Mvpd; POROK7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4597, 81726 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction