missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MVD Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98526CL1
This item is not returnable.
View return policy
Description
MVD Polyclonal antibody specifically detects MVD in Human samples. It is validated for Western Blot
Specifications
| MVD | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| EC 4.1.1.33, FP17780, MDDase, mevalonate (diphospho) decarboxylase, Mevalonate (diphospho)decarboxylase, Mevalonate pyrophosphate decarboxylase, MPDdiphosphomevalonate decarboxylase | |
| Produced in rabbits immunized with E. coli-derived Human MVD fragment. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 4597 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction