missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MVB12B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565555
This item is not returnable.
View return policy
Description
Immunogen sequence: KLIDIKDTLPV GFIPIQETVD TQEVAFRKKR LCIKFIPRDS TEAAICDIRI MGRTKQAPPQ YTFIGELNSM GIWYRM Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
Component of the ESCRT-I complex, a regulator of vesicular trafficking process. Required for the sorting of endocytic ubiquitinated cargos into multivesicular bodies.
Specifications
| MVB12B | |
| Polyclonal | |
| Unconjugated | |
| MVB12B | |
| C9orf28; ESCRT-I complex subunit MVB12B; FAM125B; family with sequence similarity 125, member B; multivesicular body subunit 12B; MVB12B; Protein FAM125B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 89853 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H7P6 | |
| MVB12B | |
| Recombinant Human MVB12B. Recombinant protein control fragment (Product #RP-106230). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction