missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MVB12A Polyclonal Antibody
GREENER_CHOICE

Product Code. 15757621 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15757621 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15757621 Supplier Invitrogen™ Supplier No. PA559772

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LGATDTAVFD VRLSGKTKTV PGYLRIGDMG GFAIWCKKAK APRPVPKPRG LSRDMQGLSL DAASQPSKGG LLERTASRLG SRASTL Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 73%.

FAM125A, also named as MVB12A and CFBP, is a component of the ESCRT-I complex. FAM125A is required for the sorting of endocytic ubiquitinated cargos into multivesicular bodies. FAM125A has three isoforms with MW 27-28 kDa and 25 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MVB12A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MVB12A
Gene Accession No. Q96EY5
Gene Alias CFBP; CIN85/CD2AP family binding protein; CIN85/CD2AP family-binding protein; ESCRT-I complex subunit MVB12A; Fam125a; family with sequence similarity 125, member A; multivesicular body subunit 12A; Mvb12a; Protein FAM125A
Gene Symbols MVB12A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MVB12A. Recombinant protein control fragment (Product #RP-99121).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93343
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.