missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUTED/BLOC1S5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24015-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a component of BLOC-1 (biogenesis of lysosome-related organelles complex 1). Components of this complex are involved in the biogenesis of organelles such as melanosomes and platelet-dense granules. A mouse model for Hermansky-Pudlak Syndrome is mutated in the murine version of this gene. Alternative splicing results in multiple transcript variants. Read-through transcription exists between this gene and the upstream EEF1E1 (eukaryotic translation elongation factor 1 epsilon 1) gene, as well as with the downstream TXNDC5 (thioredoxin domain containing 5) gene.Specifications
| MUTED/BLOC1S5 | |
| Polyclonal | |
| Unconjugated | |
| BLOC1S5 | |
| BL1S5, BLOC1S5, MU, MUTED, muted homolog (mouse), Protein Muted homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 63915 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8TDH9 | |
| BLOC1S5 | |
| MUTED/BLOC1S5 Fusion Protein Ag21226 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction