Learn More
Elma™ Mutebox XL
Product Code. 17861727
Click to view available options
:
Mutebox XL; For Use With (Equipment): Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic Med, Elmasonic EASY, Elmasonic Xtra TT
Product Code. 17861727
Brand: Elma™ 1123006001
Only null left
Add to basket
Add to basket
The Mutebox XL reduces noise of the ultrasonic cleaning unit by up to 85%.
  • Guarantees low-noise cleaning
  • Quiet fan and optimal insulation make working with ultrasonic equipment a pleasant, health-friendly and quiet experience, whether for cleaning or sample preparation
  • Fan extracts condensation water and vapors to the rear
  • Interior lighting and glass door let you keep an eye on the cleaning process at all times
  • Two independent access points ensure that the items being cleaned and the ultrasonic unit are always easily accessible
  • Made of high-quality steel and is easy to clean, ensuring hygienic operation
Specifications
Type Mutebox
For Use With (Equipment) Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic Med, Elmasonic EASY, Elmasonic Xtra TT