missing translation for 'onlineSavingsMsg'
Learn More

Mutated EGFR-IN-1, MedChemExpress

Product Code. 16448238
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448238 10 mg 10mg
16447966 100 mg 100mg
16444127 50 mg 50mg
16420688 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16448238 Supplier Medchem Express Supplier No. HY78869

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mutated EGFR-IN-1 (Osimertinib analog) is a useful intermediate for the inhibitors design for mutated EGFR, such as L858R EGFR, Exonl9 deletion activating mutant and T790M resistance mutant.

Mutated EGFR-IN-1 (Osimertinib analog) is a useful intermediate for the inhibitors design for mutated EGFR, such as L858R EGFR, Exonl9 deletion activating mutant and T790M resistance mutant.

Specifications

Chemical Name or Material Mutated EGFR-IN-1
CAS 1421372-66-8
Color Pale Purple
Purity Grade Notes Research
Molecular Formula C25H31N7O
Quantity 10 mg
Synonym Osimertinib analog
Solubility Information DMSO : ≥ 75 mg/mL (168.33 mM)
SMILES NC1=CC(NC2=NC=CC(C3=CN(C)C4=C3C=CC=C4)=N2)=C(OC)C=C1N(CCN(C)C)C
Molecular Weight (g/mol) 445.56
Formula Weight 445.56
Percent Purity 98.59%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.