missing translation for 'onlineSavingsMsg'
Learn More

Mutant IDH1 inhibitor, MedChemExpress

Product Code. 16436376
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16436376 100 mg 100mg
16415948 10 mg 10mg
16412097 50 mg 50mg
16408577 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16436376 Supplier Medchem Express Supplier No. HY13972

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mutant IDH1 inhibitor is a potent mutant IDH1 R132H inhibitor with IC50 of < 72 nM.

Mutant IDH1 inhibitor is a potent mutant IDH1 R132H inhibitor with IC50 of < 72 nM.

Specifications

Chemical Name or Material Mutant IDH1 inhibitor
Physical Form Solid
CAS 1429180-08-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C25H34N6O3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 34 mg/mL (72.87 mM)
SMILES CC(C)[C@@H](COC1=O)N1C2=NC(N[C@@H](C)C3=CC=C(CN4CCN(C(C)=O)CC4)C=C3)=NC=C2
Molecular Weight (g/mol) 466.58
Formula Weight 466.58
Percent Purity 98.69%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.