missing translation for 'onlineSavingsMsg'
Learn More

Mutant EGFR inhibitor, MedChemExpress

Product Code. 16458577
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16458577

Brand: Medchem Express HY13984

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mutant EGFR inhibitor is a potent and selective mutant EGFR inhibitor extracted from patent WO 2013014448 A1; inhibits EGFRL858R, EGFRExon 19 deletion and EGFRT790M.

Mutant EGFR inhibitor is a potent and selective mutant EGFR inhibitor extracted from patent WO 2013014448 A1

Specifications

Chemical Name or Material Mutant EGFR inhibitor
CAS 1421373-62-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H30ClN7O2
Quantity 5 mg
Solubility Information DMSO : ≥ 50 mg/mL (96.15 mM)
SMILES C=CC(NC1=CC(NC2=NC=C(Cl)C(C3=CNC4=C3C=CC=C4)=N2)=C(OC)C=C1N(CCN(C)C)C)=O
Molecular Weight (g/mol) 520.03
Formula Weight 520.03
Percent Purity 99.1%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.