missing translation for 'onlineSavingsMsg'
Learn More

MUT056399, MedChemExpress

Product Code. 16499947
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499947 5 mg 5mg
16407478 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499947 Supplier Medchem Express Supplier No. HY18169

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MUT056399 (Fab-001) is a highly potent inhibitor of the FabI enzyme of both S. aureus and E. coli with 50% inhibitory concentration IC50s of 12 nM and 58 nM, respectively.

MUT056399 (Fab-001) is a highly potent inhibitor of the FabI enzyme of both S. aureus and E. coli with 50% inhibitory concentration IC50s of 12 nM and 58 nM, respectively.

Specifications

Chemical Name or Material MUT056399
CAS 1269055-85-7
Color White
Purity Grade Notes Research
Molecular Formula C15H13F2NO3
Quantity 5 mg
Synonym Fab-001
Solubility Information DMSO : ≥ 31 mg/mL (105.70 mM)
SMILES CCC1=CC(O)=C(OC2=CC=C(C(N)=O)C=C2F)C=C1F
Molecular Weight (g/mol) 293.27
Formula Weight 293.27
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.