missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MUT Recombinant Rabbit Monoclonal Antibody (23GB4555), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553766
This item is not returnable.
View return policy
Description
The product of this gene is a Golgi enzyme catalyzing an essential step in the conversion of oligomannose to complex N-glycans. The enzyme has the typical glycosyltransferase domains: a short N-terminal cytoplasmic domain, a hydrophobic non-cleavable signal-anchor domain, and a C-terminal catalytic domain. Mutations in this gene may lead to carbohydrate-deficient glycoprotein syndrome, type II. The coding region of this gene is intronless. Transcript variants with a spliced 5' UTR may exist, but their biological validity has not been determined.
Specifications
| MUT | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MMUT | |
| D230010K02Rik; Mcm; methylmalonyl CoA mutase; methylmalonyl Coenzyme A mutase; methylmalonyl-CoA isomerase; methylmalonyl-CoA mutase; methylmalonyl-CoA mutase, mitochondrial; methylmalonyl-Coenzyme A mutase; MMUT; MUT; mutant methylmalonyl CoA mutase; truncated methylmalonyl CoA mutase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17850, 4594, 688517 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4555 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P16332, P22033 | |
| MMUT | |
| A synthesized peptide derived from human MMUT (45-95AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction