missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17034-1-AP-20UL
This item is not returnable.
View return policy
Description
MUT encodes the mitochondrial enzyme methylmalonyl Coenzyme A mutase. In humans, the product of this gene is a vitamin B12-dependent enzyme which catalyzes the isomerization of methylmalonyl-CoA to succinyl-CoA, while in other species this enzyme may have different functions. Mutations in this gene may lead to various types of methylmalonic aciduria.Specifications
| MUT | |
| Polyclonal | |
| Unconjugated | |
| MUT | |
| MCM, Methylmalonyl CoA isomerase, MUT | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17850, 4594, 688517 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16332, P22033 | |
| Mut | |
| MUT Fusion Protein Ag10523 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction