Learn More
Invitrogen™ MUT Monoclonal Antibody (2D6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549182
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Immunogen sequence is different from the related mouse sequence by one amino acid. Positive Control - WB: human Hela whole cell, human K562 whole cell, human HEK293 whole cell, human PC-3 whole cell, human Caco-2 whole cell, human Raji whole cell, rat PC-12 whole cell, mouse NIH/3T3 whole cell. IHC: human mammary cancer tissue, human rectal cancer tissue, human rectal cancer tissue, rat intestine tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The product of this gene is a Golgi enzyme catalyzing an essential step in the conversion of oligomannose to complex N-glycans. The enzyme has the typical glycosyltransferase domains: a short N-terminal cytoplasmic domain, a hydrophobic non-cleavable signal-anchor domain, and a C-terminal catalytic domain. Mutations in this gene may lead to carbohydrate-deficient glycoprotein syndrome, type II. The coding region of this gene is intronless. Transcript variants with a spliced 5' UTR may exist, but their biological validity has not been determined.
Specifications
| MUT | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P16332, P22033 | |
| MMUT | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human MUT (33-65aa LHQQQPLHPEWAALAKKQLKGKNPEDLIWHTPE). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2D6 | |
| Unconjugated | |
| MMUT | |
| D230010K02Rik; Mcm; methylmalonyl CoA mutase; methylmalonyl Coenzyme A mutase; methylmalonyl-CoA isomerase; methylmalonyl-CoA mutase; methylmalonyl-CoA mutase, mitochondrial; methylmalonyl-Coenzyme A mutase; MMUT; MUT; mutant methylmalonyl CoA mutase; truncated methylmalonyl CoA mutase | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 17850, 4594, 688517 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.