Learn More
Invitrogen™ MUSK Recombinant Rabbit Monoclonal Antibody (PSH05-68)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552463
Description
Positive Control: Mouse skeletal muscle tissue lysate, rat skeletal muscle tissue lysate, human skeletal muscle tissue, mouse skeletal muscle tissue, rat skeletal muscle tissue. Sequence Similarities: 100% Mouse/Rat. Subcellular Location: Postsynaptic cell membrane. Predicted band size: 97 kDa.
MUSK plays an important and selective role in many aspects of skeletal muscle development and function through the initial step of postsynaptic differentiation and formation of end-plate bands in cooperation with Rapsyn. Communication between cells is often mediated by receptors on the surface of the target cell that are triggered by specific ligands released by other cells. Receptor tyrosine kinase activity is strictly regulated by the binding of the appropriate ligand to the extracellular active site of the receptor. MuSK has been shown to be part of the agrin signaling complex and plays an important role in the development and architectural maintenance of the neuromuscular junction.
Specifications
| MUSK | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O15146, Q61006, Q62838 | |
| MUSK | |
| Recombinant protein within human musk aa 1-516/869. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| PSH05-68 | |
| Unconjugated | |
| MUSK | |
| CMS9; FADS; Mdk4; MGC126323; MGC126324; Mlk; muscle associated receptor tyrosine kinase; muscle localized kinase 2; muscle specific kinase (neural fold/somite kinase 1); muscle, skeletal receptor tyrosine protein kinase; Muscle, skeletal receptor tyrosine-protein kinase; muscle, skeletal, receptor tyrosine kinase; muscle-specific kinase receptor; muscle-specific protein kinase secretory isoform; muscle-specific tyrosine kinase receptor MuSK; muscle-specific tyrosine protein kinase receptor; Muscle-specific tyrosine-protein kinase receptor; MuSK; Nsk1; Nsk2; Nsk3; RP23-170A3.1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18198, 4593, 81725 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.