missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MuSK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-5619
This item is not returnable.
View return policy
Description
MuSK Polyclonal specifically detects MuSK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| MuSK | |
| Polyclonal | |
| Unconjugated | |
| PBS, 1.0 mg/ml BSA and 30% Glycerol with 0.05% Sodium Azide | |
| MUSK | |
| Recombinant protein containing residues 210-304 from the extracellular domain of rat MuSK | |
| 200 μg | |
| Protein Kinase, Signal Transduction | |
| 4593 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:20 - 1:100, Immunocytochemistry/Immunofluorescence 1:20 - 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20 - 1:100 | |
| EC 2.7.10, EC 2.7.10.1, MGC126323, MGC126324, muscle, skeletal, receptor tyrosine kinase, MuSK, skeletal receptor tyrosine-protein kinase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects MUSK from mouse and rat tissue samples as well as COS7 cells transfected with rat MUSK. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction