missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscleblind-like 1 Rabbit anti-Human, Clone: 7N6W3, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Muscleblind-like 1 |
|---|---|
| Clone | 7N6W3 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18359885
|
Novus Biologicals
NBP3-16574-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18342504
|
Novus Biologicals
NBP3-16574-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Muscleblind-like 1 Monoclonal antibody specifically detects Muscleblind-like 1 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Muscleblind-like 1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, DNA replication Transcription Translation and Splicing | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4154 | |
| IgG | |
| Affinity purified |
| 7N6W3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp686P06174, EXP40, EXP42, EXPKIAA0428EXP35, MBNL, muscleblind (Drosophila)-like, muscleblind-like (Drosophila), muscleblind-like protein 1, Triplet-expansion RNA-binding protein | |
| A synthetic peptide corresponding to a sequence within amino acids 289-388 of human Muscleblind-like 1 (Q9NR56). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title