missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscle Phosphofructokinase/PFKM/PFK-1 Antibody, Biotin, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Muscle Phosphofructokinase/PFKM/PFK-1 Polyclonal antibody specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Muscle Phosphofructokinase/PFKM/PFK-1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | 6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 730 to 780 of human Phosphofructokinase, Muscle using the numbering given in entry NP_000280.1 (GeneID 5213). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?