missing translation for 'onlineSavingsMsg'
Learn More

Muscle Phosphofructokinase/PFKM/PFK-1 Antibody (842735) [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30738226 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30738226 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738226 Supplier Novus Biologicals Supplier No. FAB7687J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Muscle Phosphofructokinase/PFKM/PFK-1 Monoclonal antibody specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Muscle Phosphofructokinase/PFKM/PFK-1
Applications Western Blot
Classification Monoclonal
Clone 842735
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias 6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase
Host Species Mouse
Immunogen E. coli-derived recombinant human Muscle Phosphofructokinase/PFKM, Asn674-Val780, Accession # P08237
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5213
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.