missing translation for 'onlineSavingsMsg'
Learn More

Muscle Phosphofructokinase/PFKM/PFK-1 Antibody (2F11), HRP, Novus Biologicals™

Product Code. 18419431 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419431 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419431 Supplier Novus Biologicals Supplier No. NBP222615H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Muscle Phosphofructokinase/PFKM/PFK-1 Monoclonal antibody specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Muscle Phosphofructokinase/PFKM/PFK-1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2F11
Conjugate HRP
Formulation PBS
Gene Alias 6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase
Host Species Mouse
Immunogen Recombinant human Muscle Phosphofructokinase/PFKM/PFK-1(1-780aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5213
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.